Anti-GSTM1 Mouse Monoclonal Antibody [clone: 1H4A4]

Supplier: Genetex
GTX60526
10168-950EA 625.73 USD
10168-950
Anti-GSTM1 Mouse Monoclonal Antibody [clone: 1H4A4]
Antibodies
Mouse Monoclonal Antibody [1H4A4] to GSTM1

Type: Primary
Antigen: GSTM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR