Anti-Thra Rabbit Polyclonal Antibody

Supplier: Genetex
GTX82858
10168-852EA 625.36 USD
10168-852
Anti-Thra Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Thyroid Hormone Receptor alpha-2

Type: Primary
Antigen: Thra
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR