Anti-Aanat Rabbit Polyclonal Antibody

Supplier: Genetex
GTX23506
10168-690EA 624.92 USD
10168-690
Anti-Aanat Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Serotonin N-AT

Type: Primary
Antigen: Aanat
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR