Anti-FZD4 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX71851
10168-310EA 624.58 USD
10168-310
Anti-FZD4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to Human Frizzled - 4 ( FZD4 )

Type: Primary
Antigen: FZD4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR