Anti-IgG Mouse Monoclonal Antibody [clone: GT1056] (HRP (Horseradish Peroxidase))

Supplier: Genetex
GTX628995-01
10168-052EA 324.08 USD
10168-052
Anti-IgG Mouse Monoclonal Antibody [clone: GT1056] (HRP (Horseradish Peroxidase))
Antibodies
HRP-conjugated Mouse anti-Goat IgG (Heavy chain) Monoclonal antibody [GT1056]

Type: Secondary
Antigen: IgG
Clonality: Monoclonal
Clone: GT1056
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR