Anti-RAB9 Mouse Monoclonal Antibody [clone: Mab9]

Supplier: Genetex
GTX22810
10167-998EA 625.18 USD
10167-998
Anti-RAB9 Mouse Monoclonal Antibody [clone: Mab9]
Antibodies
Mouse monoclonal antibody [Mab9] to Rab9

Type: Primary
Antigen: Ras-related protein Rab-9A
Clonality: Monoclonal
Clone: Mab9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog, Monkey, Rat, Hamster, Primates, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR