Anti-Tcp1 Rat Monoclonal Antibody [clone: 91A]

Supplier: Genetex
GTX22797
10167-978EA 626.93 USD
10167-978
Anti-Tcp1 Rat Monoclonal Antibody [clone: 91A]
Antibodies
Rat monoclonal antibody [91A] to TCP1

Type: Primary
Antigen: Tcp1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Chicken, Dog, Rat, Yeast, Hamster, Primates
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR