Anti-PITX2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX129141
10167-710EA 668.55 USD
10167-710
Anti-PITX2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PITX2 (paired-like homeodomain 2)

Type: Primary
Antigen: PITX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR