Anti-STC1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX129092
10167-698EA 588.68 USD
10167-698
Anti-STC1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Stanniocalcin 1 (stanniocalcin 1)

Type: Primary
Antigen: STC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR