Anti-MX1 Mouse Monoclonal Antibody [clone: GT4812]

Supplier: Genetex
GTX629738
10167-454EA 668.55 USD
10167-454
Anti-MX1 Mouse Monoclonal Antibody [clone: GT4812]
Antibodies
Mouse Monoclonal antibody [GT4812] to MX1 (myxovirus (influenza virus) resistance 1, interferon-inducible protein p78 (mouse))

Type: Primary
Antigen: MX1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR