Anti-SS18 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX129428
10167-032EA 588.68 USD
10167-032
Anti-SS18 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SS18 (synovial sarcoma translocation, chromosome 18)

Type: Primary
Antigen: SS18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR