Prepacked Solid Load Cartridges for RediSep® Rf column, Teledyne ISCO
Supplier: Teledyne
693873241
101663-932PK 373.85 USD
101663-932
Prepacked Solid Load Cartridges for RediSep® Rf column, Teledyne ISCO
Chromatography Columns
Solid sample loading is a useful technique to load samples to be purified with a RediSep® Rf column, particularly in the case of low-solubility samples. All CombiFlash systems are designed for either solid or liquid sample loading.
  • Used with a solid load cartridge cap
  • Adjustable cap minimizes void volumes
  • Accurate gradients
Several sizes of sample load cartridges (5, 25, 65, and 270 gram nominal sorbent capacity) allow loading of sample amounts from milligrams to tens of grams.
Delivery information: Available either empty (to fill with your own sorbent) or pre-filled with silica gel or reversed-phase C18 material.