Anti-LCMT1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX120404
10166-270EA 588.68 USD
10166-270
Anti-LCMT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to LCMT1 (leucine carboxyl methyltransferase 1)

Type: Primary
Antigen: LCMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR