Anti-CBX6 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX119535
10165-978EA 588.68 USD
10165-978
Anti-CBX6 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CBX6 (chromobox homolog 6)

Type: Primary
Antigen: CBX6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR