Anti-CHRDL1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX117884
10165-946EA 588.52 USD
10165-946
Anti-CHRDL1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CHRDL1 (chordin-like 1)

Type: Primary
Antigen: CHRDL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR