Anti-PCOLCE2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX103410
10165-928EA 588.68 USD
10165-928
Anti-PCOLCE2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PCOLCE2 (procollagen C-endopeptidase enhancer 2)

Type: Primary
Antigen: PCOLCE2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR