Anti-Pcsk4 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX12306
10165-444EA 640.76 USD
10165-444
Anti-Pcsk4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PCSK4

Type: Primary
Antigen: Pcsk4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR