Anti-Tram2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX12287
10165-418EA 640.76 USD
10165-418
Anti-Tram2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TRAM2

Type: Primary
Antigen: Tram2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR