Anti-Ibsp Rabbit Polyclonal Antibody

Supplier: Genetex
GTX12155
10165-366EA 656.47 USD
10165-366
Anti-Ibsp Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Bone Sialoprotein

Type: Primary
Antigen: Ibsp
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR