Anti-Prdx3 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX12108
10165-354EA 626.01 USD
10165-354
Anti-Prdx3 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PRDX3

Type: Primary
Antigen: Prdx3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR