Anti-TSPAN13 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX52155
10165-014EA 625.36 USD
10165-014
Anti-TSPAN13 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TSPAN13

Type: Primary
Antigen: TSPAN13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR