Hard-Shell® 96-Well Semi-Skirted PCR Plates, High-Profile, Bio-Rad

Supplier: Bio-Rad
HSS9601
101647-160EA 215.38 USD
101647-160
Hard-Shell® 96-Well Semi-Skirted PCR Plates, High-Profile, Bio-Rad
Microplates PCR Plates
Hard-Shell 96-Well Semi-Skirted PCR Plates are designed to withstand the stresses of thermal cycling and robotic handling

  • Reaction volumes of 5–125 µL (350 µL maximum)
  • Black lettering for easy well identification
  • Free of DNase, RNase and human DNA
  • White-well option for increased fluorescent signal strength
  • Color-coded skirt option with clear wells
  • Low-cost, user-readable bar code option for convenient database tracking

The raised rims around each well allow tight sealing using different procedures including pressure-, adhesive-, and heat-sealing methods. The high-profile (20.75 mm) wells fit most thermal cyclers, real-time PCR detection systems, and DNA sequencers.

The patented two-component design provides stability and flatness, allowing precise positioning for automation. The skirt and deck are molded from a rigid thermostable polymer to eliminate warping and shrinkage, and the thin-wall wells are molded of virgin polypropylene with low DNA binding. The half-height skirt provides improved robotic handling and a labeling surface.

Compatible with: CFX96™, 1000-Series, DNA Engine Opticon® Series, Chromo4™, PTC-100®, DNA Engine® Series, Applied Biosystems standard systems (7300, 7500, 9700, 2070, Veriti, etc.), Applied Biosystems Sanger sequencers (3700, 3730, etc.), Eppendorf Mastercyclers.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR