Microseal® 'B' Adhesive Seals, Optical, Bio-Rad
Supplier: Bio-Rad
ADR3296
101647-200EA 231 USD
101647-200 101646-984 101647-208
Microseal® 'B' Adhesive Seals, Optical, Bio-Rad
Seals Microplate Seals
Microseal® 'B' Adhesive Seals are designed for use with thin-wall PCR plates. The aggressive adhesive layer ensures secure sample storage or transport before or after cycling. Perforated end tabs can be removed when using these seals on semi-skirted PCR plates or with automated plate handlers.
The clear polyester film of Microseal® 'B' Adhesive Seals allows inspection of sample wells and effective light transmission during real-time PCR and other optical assays.
For 96-well plates, the optional compression pad can enhance seal integrity during thermal cycling.
The adhesive is effective from –40 to 110°C. The seals are free of DNase, RNase, and human DNA.