BioFrac™ Fraction Collector, Bio-Rad
Supplier: Bio-Rad
7410002
101646-916EA 11624.62 USD
101646-916
BioFrac™ Fraction Collector, Bio-Rad
Fraction Collectors
The BioFrac™ Fraction Collector has the flexibility for both analytical and preparative chromatography applications, is easy to program, and can be used for basic or complex fraction collection schemes at flow rates ≤100 ml/min.
  • Range of collection modes
  • Collection by peak detection, time windows (up to 20), or a combination of both
  • Numerous flatpack off-the-shelf racks to accommodate tubes (12–20 mm dia), microtubes (0.5, 1.5, or 2.0 ml), or scintillation vials (16 or 30 mm dia)
  • Drop arm movement in column, row, or serpentine pattern option for microplates or Titertube® Micro Test Tubes
  • Dispenser arm adjusts to tube heights ≤150 mm
  • Multi-run feature allows overlay of fractions
  • Optional BioFrac™ Ice Bath/Microplate Rack with tube grips that hold tubes firmly in any position; allows cooling of 13 mm tubes or serves as a rack for microplates and tubes meeting SBS standards
  • Optional BioFrac™ Prep-20 Preparative Rack with 20 ports for fractionation into 20 vessels of any size up to carboys
  • Optional Microplate Drophead Kit for precise collection of small volumes into microplates
  • Polyurethane waste outlet tube
  • Polypropylene case and carousel
  • Hard-coated polycarbonate front panel
The numerous collection rack options include both custom and off-the-shelf collection racks. Custom racks provide a wide range of collection capability, from drops to liters. Off-the-shelf racks extend the versatility of collection schemes and provide cost-effective storage of samples. The off-the-shelf racks are autoclavable, easily assembled, and lie flat, taking little storage space.
Medium-contacting material: All non-metal, including Tefzel, PEEK
Certifications: Meets IEC 61010 and CSA 22.2 certification.