Quick Start™ Bradford Protein Assay and Protein Standards, Bio-Rad
Supplier: 0000013515
5000209 5000204 5000205 5000207 5000202 5000201 5000206 5000208
101646-090EA 318.46 USD
101646-090 101646-080 101646-078 101646-082 101646-086 101646-076 101646-074 101646-084 101646-088
Quick Start™ Bradford Protein Assay and Protein Standards, Bio-Rad
Assays Protein Assays
The Quick Start™ Bradford Protein Assay is a simple, accurate assay for determining the concentration of protein in solution, with a linear range of 1.25–25 µg/mL.
The assay kits contain ready-to-use dye reagent with either bovine serum albumin (BSA) or bovine γ-globulin standards. For either standard, choose between seven prediluted concentrations (0.125, 0.25, 0.5, 0.75, 1.0, 1.5, and 2.0 mg/ml), or making your own standard series from 2mg/ml stock solutions. The Quick Start™ Bradford Dye Reagent and Protein Standards are also available separately.