Criterion™ XT Bis-Tris Precast Gels, Midi Protein Gels, Bio-Rad
Supplier: Bio-Rad
3450118 3450123 3450115 3450127 3450111 3450121 3450126 3450124 3450113 3450112 3450117 3450119 3450125
101645-912EA 29.23 USD
101645-912 101645-920 101645-908 101645-928 101645-902 101645-916 101645-918 101645-926 101645-922 101645-906 101645-904 101645-910 101645-914 101645-924
Criterion™ XT Bis-Tris Precast Gels, Midi Protein Gels, Bio-Rad
Electrophoresis Gels Precast Gels
Criterion™ XT Bis-Tris Precast Gels are formulated using a Bis-Tris buffer system (pH 6.4) and can be run with either XT MOPS or XT MES running buffers
  • Useful for screening new samples and evaluating sample preparation conditions
  • Laemmli-type separation
  • Run 2-D gels in less than a day
  • Near-neutral pH delays acrylamide hydrolysis significantly longer than Laemmli buffers
  • Very stable, with a 12 month shelf life at room temperature
The ability to use two different buffer systems provides the flexibility to obtain different separation profiles with only one type of gel. XT MOPS buffer system is commonly used for mid-sized proteins and XT MES buffer system for smaller proteins.
Ordering information: Criterion™ XT Bis-Tris Precast Gels are packaged and sold in individual units: 1–2 gels can be run in Bio-Rad’s Criterion™ Cell, and 1–12 gels in the high-throughput Criterion™ Dodeca™ Cell.