EZ Micro and Standard Micro Test Tubes, Bio-Rad
Supplier: Bio-Rad
2239500
101645-698EA 36.92 USD
101645-698 101645-694
EZ Micro and Standard Micro Test Tubes, Bio-Rad
Tubes Microtubes
To fit centrifuges, 2 ml tubes have thinner walls than standard tubes and are intended for short runs at less than 13000 ×g. Heavy samples, high g forces, long runs, or centrifuges that radiate excess heat may damage these tubes.
  • Suitable for general-purpose benchtop centrifuge use
  • Constructed with sturdy, uniform walls that easily withstand up to 13000 ×g
  • Autoclavable to 120 °C and freezable to –80 °C
  • Available in 2 ml, 1.5 ml, and 500 µl sizes
Separate polyethylene caps for the 1.5 ml capless tubes have a knurled top for easy handling.