iQ™ 96-Well Real-Time PCR Plates, High Profile, Bio-Rad
Supplier: Bio-Rad
2239441
101645-684EA 486.15 USD
101645-684
iQ™ 96-Well Real-Time PCR Plates, High Profile, Bio-Rad
Microplates PCR Plates
iQ™ 96-Well Real-Time PCR Plates are high-profile, semi-skirted plates, compatible with many real-time PCR detection systems including, the Bio-Rad 1000 series, Applied Biosystems standard systems, the Eppendorf Mastercycler series and Agilent (Stratagene) MX series.
  • Half-height skirt adds stiffness and a labeling surface
  • Recommended volumes of 5–125 µL (330 µL maximum)
  • Perforated every three columns for easy setup of triplicate reactions
  • Free of DNase, RNase, and human DNA