iScript RT-qPCR Sample Preparation Reagent, Bio-Rad
Supplier: 0000013515
1708899 1708898
101645-420EA 1156.92 USD
101645-420 101645-418
iScript RT-qPCR Sample Preparation Reagent, Bio-Rad
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
iScript™ RT-qPCR sample preparation reagent provides a simple and rapid procedure (<10 min) for isolating total RNA from cultured cells to use for RT-qPCR. This reagent delivers efficient cell lysis, RNA stabilization, and removal of genomic DNA for sensitive qPCR without traditional RNA sample preparation.
* Have ease of use and simplicity — one-buffer formulation contains all of the necessary components to perform cell lysis, RNA stabilization, and genomic DNA removal in 5–10 min
Key Features and Benefits:
* Have ease of use and simplicity — one-buffer formulation contains all of the necessary components to perform cell lysis, RNA stabilization, and genomic DNA removal in 5–10 min
* Benefit from superior compatibility with qPCR directly from cells — sensitive detection of high-, medium-, and low-copy gene targets directly from cell lysates using real-time qPCR when combined with iScript reverse transcription and qPCR reagents
* Maximize data from very few cells — reagent enables multiplex real-time detection of up to 4 targets from as few as 10 cells
* Extend isolated RNA shelf-life — isolated RNA is stable for up to 6 months when frozen
* Use with different cell types — compatible with both suspension and adherent cell cultures
Packaging: Contains RNase inhibitors and RNA stabilizers