Tween® 20 (Polysorbate) 100% Nonionic Detergent, Enzyme Immunoassay Grade

Supplier: BIO-RAD LABORATORIES MS

1706531
101645-278EA 118.46 USD
101645-278
Tween® 20 (Polysorbate) 100% Nonionic Detergent, Enzyme Immunoassay Grade
Tween® 20 (Polysorbate)

Choose 100% Tween 20 for applications including protein extractions, and blocking and washing of western blots and ELISAs.

Formula: C₅₈H₁₁₄O₂₆
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR