HRP and AP Antibody Conjugates, Bio-Rad
Supplier: 0000013515
1706521 1706528 1706520 1706533 1705047 1706522 1705046 1706425 1706516 1721050 1706518 1706515
101645-270EA 389.23 USD
101645-270 101645-276 101645-268 101645-282 101645-224 101645-272 101645-222 101645-242 101645-264 101645-448 101645-266 101645-262
HRP and AP Antibody Conjugates, Bio-Rad
Electrophoresis Reagents Western Blotting Reagents
Bio-Rad Horseradish Peroxidase (HRP) and Alkaline Phosphatase (AP) Secondary Antibody Conjugates are engineered for use with blot detection protocols including colorimetric and chemiluminescent.
Double affinity-purified blotting-grade antibodies are isolated by affinity chromatography, and further purified by cross-adsorption against an unrelated species to eliminate nonspecific immunoglobulins.
High titer blotting-grade antibody conjugates increase assay sensitivity; greater working dilutions (1:3,000) decrease background and increase the signal-to-noise ratio.