Bio-Lyte 3/10 ampholyte Broad pH Range (~3.5–9.5), 100x
Supplier: BIO-RAD LABORATORIES MS
1632094
101644-036EA 118.46 USD
101644-036
Bio-Lyte 3/10 ampholyte Broad pH Range (~3.5–9.5), 100x
Bio-Lyte
Bio-Lyte carrier ampholytes, supplied as clear aqueous solutions, are blended to give a complete range of isoelectric points for reproducible, linear pH gradients. For use in IEF and with ReadyStrip™ IPG strips
* Wide-range Bio-Lyte 3/10 ampholytes have a working pH range of 3.5–9.5.