ReadyPrep Protein Extraction Kit (Cytoplasmic/Nuclear), Bio-Rad
Supplier: BIO-RAD LABORATORIES MS
1632089
101644-026EA 986.15 USD
101644-026
ReadyPrep Protein Extraction Kit (Cytoplasmic/Nuclear), Bio-Rad
Protein Extraction Protein Extraction Kits
The ReadyPrep protein extraction kit (cytoplasmic/nuclear) prepares fractions enriched in cytoplasmic and nuclear proteins from eukaryotic samples, such as cultured cells and tissues.
The ReadyPrep protein extraction kit (cytoplasmic/nuclear):
* Employs a specially formulated buffer and differential centrifugation to isolate intact nuclei (Dignam et al. 1983, Zerivitz and Akusjarvi 1989)
* Contains a strongly chaotropic extraction buffer to efficiently solubilize nuclear proteins
* Includes the components of the ReadyPrep 2-D cleanup kit to remove compounds from the extraction buffer that may interfere with IEF
* Employs a specially formulated buffer and differential centrifugation to isolate intact nuclei (Dignam et al
References
- Dignam JD et al. (1983). Accurate transcription initiation by RNA polymerase II in a soluble extract from isolated mammalian nuclei. Nucleic Acid Res 11, 1475–1489.
- Zerivitz K and Akusjarvi G (1989). An improved nuclear extract preparation method. Gene Anal Tech 6, 101–109.