Immun-Blot® PVDF Membrane, Bio-Rad
Supplier: Bio-Rad
1620175 1620176 1620174 1620239 1620219 1620255
101643-834EA 272.31 USD
101643-834 101643-898 101643-836 101643-832 101643-900 101643-882 101643-884 101643-910
Immun-Blot® PVDF Membrane, Bio-Rad
Transfer Membranes
Immun-Blot PVDF Membrane is engineered for western blots with chemiluminescent and colorimetric detection
PVDF membrane retains proteins strongly during protein transfer, and has nonspecific protein binding during processing giving low backgrounds and high-sensitivity. High mechanical strength permits reprobing of individual blots.
Ordering information: Immun-Blot PVDF Membranes are available in three formats; precut membrane/blot absorbent filter paper sandwiches, packs of precut membrane-only sheets and rolls.