Sequi-Blot™ PVDF Membrane, Bio-Rad
Supplier: Bio-Rad
1620184
101643-842EA 203 USD
101643-842 101643-894 101643-840 101643-846 101643-896 101643-848 101643-844
Sequi-Blot™ PVDF Membrane, Bio-Rad
Transfer Membranes
Sequi-Blot™ PVDF membrane is ideal for protein sequencing: It retains target protein very strongly during protein transfer and provides the binding capacity to sequence even low-abundance samples.
* Mechanical strength and chemical resistance provide ideal substrate for solvents involved in protein sequencing
Features and Benefits:
* Mechanical strength and chemical resistance provide ideal substrate for solvents involved in protein sequencing
* Available in three formats:
- Convenient precut membrane/blot absorbent filter paper sandwiches
- Packs of precut membrane-only sheets
- Economical rolls
Applications:
* Protein transfer from SDS-PAGE gels to membrane
* Protein sequencing of target protein