Tween® 20 (Polysorbate) 10% in aqueous solution

Supplier: BIO-RAD LABORATORIES MS

1610781
101643-416EA 80 USD
101643-416
Tween® 20 (Polysorbate) 10% in aqueous solution
Tween® 20 (Polysorbate)
Formula: C₅₈H₁₁₄O₂₆
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR