Sodium dodecyl sulfate (SDS) 200 g/l in aqueous solution

Supplier: BIO-RAD LABORATORIES MS

1610418
101643-244EA 272.31 USD
101643-244
Sodium dodecyl sulfate (SDS) 200 g/l in aqueous solution
Sodium dodecyl sulphate (SDS)
Formula: CH₃(CH₂)₁₁OSO₃Na
Density: 1…1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR