Coomassie® brilliant blue R-250

Supplier: BIO-RAD LABORATORIES MS

1610400
101643-234EA 47.69 USD
101643-234
Coomassie® brilliant blue R-250
Coomassie® Brilliant Blue R-250

Coomassie Brilliant Blue R-250 protein stain powder

Formula: C₄₅H₄₄N₃NaO₇S₂
MDL Number: MFCD00041762
CAS Number: 6104-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR