pSNAPf Vector, New England Biolabs
Supplier: New England Biolabs (NEB)
N9183S
101641-580EA 200 USD
101641-580
pSNAPf Vector, New England Biolabs
Vectors, Plasmids and Libraries
pSNAPf Vector is a mammalian expression plasmid intended for the cloning and stable or transient expression of SNAP-tag® protein fusions in mammalian cells
This plasmid encodes SNAPf, a SNAP-tag protein, which is expressed under control of the CMV promoter. The expression vector has an IRES (internal ribosome entry site) and a neomycin resistance gene downstream of the SNAPf for the efficient selection of stable transfectants. pSNAPf Vector contains two multiple cloning sites to allow cloning of the fusion partner as a fusion to the N- or C-terminus of the SNAPf.