RNA 5’ Pyrophosphohydrolase
Supplier: New England Biolabs (NEB)
M0356S
101641-568EA 112 USD
101641-568
RNA 5’ Pyrophosphohydrolase
Enzymes
The bacterial RNA 5´ Pyrophosphohydrolase (RppH) removes pyrophosphate from the 5´ end of triphosphorylated RNA to leave a 5´ monophosphate RNA
The RppH protein was also known as NudH/YgdP which can split diadenosine penta-phosphate to ADP and ATP.