Anti-SNAP-TAG® Polyclonal Antibody
Supplier: New England Biolabs (NEB)
P9310S
101641-074EA 327 USD
101641-074
Anti-SNAP-TAG® Polyclonal Antibody
Antibodies
The Anti-SNAP-tag Antibody (Polyclonal) can be used in Western blots with SNAP-tag and CLIP-tag proteins. Polyclonal antibodies are produced from the immunization of rabbit with purified recombinant SNAP-tag protein and affinity purified using Protein A Affinity Chromatography resin.