5-methyl-dCTP, New England Biolabs
Supplier: New England Biolabs (NEB)
N0356S
101641-034EA 86 USD
101641-034
5-methyl-dCTP, New England Biolabs
Nucleic Acid Reagents Nucleotides
5-methyl-dCTP (2'-deoxy-5-methylcytidine 5'-triphosphate) is supplied as a 10 mM solution at pH 7
Nucleotide concentration is determined by measurements of absorbance at 260 nm.