Magnesium Chloride (MgCl₂) Solution, New England Biolabs
Supplier: New England Biolabs (NEB)
B9021S
101640-780EA 28 USD
101640-780
Magnesium Chloride (MgCl₂) Solution, New England Biolabs
Buffers Biological Buffers
25 mM MgCl2 for use in conjunction with polymerase reaction buffers.