Anti-TNFRSF13B Rabbit Polyclonal Antibody

Supplier: Genetex
GTX51554
10164-776EA 625.36 USD
10164-776
Anti-TNFRSF13B Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TNFRSF13B

Type: Primary
Antigen: TNFRSF13B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR