Anti-TREML1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX51550
10164-768EA 625.36 USD
10164-768
Anti-TREML1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TREML1

Type: Primary
Antigen: TREML1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR