Anti-MYOCD Rabbit Polyclonal Antibody

Supplier: Genetex
GTX13842
10163-376EA 625.35 USD
10163-376
Anti-MYOCD Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Myocardin

Type: Primary
Antigen: MYOCD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR