Anti-Ascl2 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX44645
10163-174EA 625.36 USD
10163-174
Anti-Ascl2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ASCL2

Type: Primary
Antigen: Ascl2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Dog, Pig, Rat, Guinea Pig, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR