JACKSON SAFETY® V80 MONOGOGGLE™ XTR OTG Goggle, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Ansell Healthcare
18624
10160-704CS 180.59 USD
10160-704 10160-706 10160-708
JACKSON SAFETY® V80 MONOGOGGLE™ XTR OTG Goggle, KIMBERLY-CLARK PROFESSIONAL®
Goggles
The JACKSON SAFETY® V80 Monogoggle XTR OTG Goggle protects against splashes, while providing increased visibility due to its cylindrical lens.
A soft, pliable frame conforms to the face for comfort and is easy to clean. Indirect ventilation.
Can be worn over most prescription eyeglasses.
Certifications: ANSI Z87.1-2010 Standard.