KIMTECH PURE® A5 Cleanroom Integrated Hood and Mask, Kimberly-Clark
Supplier: Ansell Healthcare
49828
10160-684CS 886.99 USD
10160-684
KIMTECH PURE® A5 Cleanroom Integrated Hood and Mask, Kimberly-Clark
Disposable Garments
In cleanroom environments, people are the number one source of contamination, and the garments they wear can make a big difference
  • Breathable SMS fabric
  • Clean processed
  • Bound, tightly sewn seams offer added protection and strengthen barriers
  • Stretch fit elastic
  • Combines two donning steps into one
  • Pull down ties in back to assist in the gowning process
  • Bulk packaged for convenience and waste reduction
  • Recyclable through the RightCycle™ by Kimberly-Clark Professional®
The KIMTECH PURE® A5 Cleanroom Integrated Hood and Mask is designed to protect cleanroom processes by providing comfortable and sustainable protection.