SENSATION® Powder free Examination Gloves, Microflex®, Ansell
Supplier: Ansell Healthcare
N730 N731 N732 N733 N734
10159-898BX 22.44 USD
10159-898 10159-900 10159-902 10159-904 10159-906
SENSATION® Powder free Examination Gloves, Microflex®, Ansell
Gloves
Powder-free, blue-colored examination gloves with enhanced comfort and tactile sensitivity.
  • Gloves are nonsterile and latex-free
  • Powder-free
  • Ambidextrous
  • Beaded cuffs
  • Disposable nitrile goves
Microflex® Sensation® N73 powder-free examination gloves provide exceptional fit with textured fingertips for an excellent grip for use with small instruments. FDA cleared 'Low Dermatitis' claim, which means Sensation® gloves have been clinically shown to reduce incidence of the redness and itching associated with contact dermatitis.